TIGIT Monoclonal Antibody (MBSA43), eFluor™ 450, eBioscience™ - FAQs
TIGIT Monoclonal Antibody (MBSA43), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TIGIT Monoclonal Antibody (MBSA43), eFluor™ 450, eBioscience™ - FAQs (48-9500-42, 48-9500-41)