Ly-6G Monoclonal Antibody (1A8-Ly6g), eFluor™ 450, eBioscience™ - FAQs
Ly-6G Monoclonal Antibody (1A8-Ly6g), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Ly-6G Monoclonal Antibody (1A8-Ly6g), eFluor™ 450, eBioscience™ - FAQs (48-9668-80, 48-9668-82)