CD303a Monoclonal Antibody (201A), eFluor™ 450, eBioscience™ - FAQs
CD303a Monoclonal Antibody (201A), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD303a Monoclonal Antibody (201A), eFluor™ 450, eBioscience™ - FAQs (48-9818-42, 48-9818-41)