IRF4 Monoclonal Antibody (3E4), eFluor™ 450, eBioscience™ - FAQs
IRF4 Monoclonal Antibody (3E4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IRF4 Monoclonal Antibody (3E4), eFluor™ 450, eBioscience™ - FAQs (48-9858-82, 48-9858-80)