CD235a (Glycophorin A) Monoclonal Antibody (6A7M), eFluor™ 450, eBioscience™ - FAQs
CD235a (Glycophorin A) Monoclonal Antibody (6A7M), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD235a (Glycophorin A) Monoclonal Antibody (6A7M), eFluor™ 450, eBioscience™ - FAQs (48-9884-42)