GARP Monoclonal Antibody (YGIC86), eFluor™ 450, eBioscience™ - FAQs
GARP Monoclonal Antibody (YGIC86), eFluor™ 450, eBioscience™ - FAQs
View additional product information for GARP Monoclonal Antibody (YGIC86), eFluor™ 450, eBioscience™ - FAQs (48-9891-82)