CD278 (ICOS) Monoclonal Antibody (ISA-3), eFluor™ 450, eBioscience™ - FAQs
CD278 (ICOS) Monoclonal Antibody (ISA-3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD278 (ICOS) Monoclonal Antibody (ISA-3), eFluor™ 450, eBioscience™ - FAQs (48-9948-42, 48-9948-41)