HLA-DR Monoclonal Antibody (L243), eFluor™ 450, eBioscience™ - FAQs
View additional product information for HLA-DR Monoclonal Antibody (L243), eFluor™ 450, eBioscience™ - FAQs (48-9952-42, 48-9952-41)