HLA-DR Monoclonal Antibody (LN3), eFluor™ 450, eBioscience™ - FAQs
HLA-DR Monoclonal Antibody (LN3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for HLA-DR Monoclonal Antibody (LN3), eFluor™ 450, eBioscience™ - FAQs (48-9956-42)