Kappa Light Chain Monoclonal Antibody (TB28-2), eFluor™ 450, eBioscience™ - FAQs
Kappa Light Chain Monoclonal Antibody (TB28-2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Kappa Light Chain Monoclonal Antibody (TB28-2), eFluor™ 450, eBioscience™ - FAQs (48-9970-42, 48-9970-41)