CD279 (PD-1) Monoclonal Antibody (RMP1-30), eFluor™ 450, eBioscience™ - FAQs
CD279 (PD-1) Monoclonal Antibody (RMP1-30), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (RMP1-30), eFluor™ 450, eBioscience™ - FAQs (48-9981-80, 48-9981-82)