CD184 (CXCR4) Monoclonal Antibody (2B11), eFluor™ 450, eBioscience™ - FAQs
CD184 (CXCR4) Monoclonal Antibody (2B11), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD184 (CXCR4) Monoclonal Antibody (2B11), eFluor™ 450, eBioscience™ - FAQs (48-9991-80, 48-9991-82)