Perforin Monoclonal Antibody (dG9 (delta G9)), eFluor™ 450, eBioscience™ - FAQs
Perforin Monoclonal Antibody (dG9 (delta G9)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Perforin Monoclonal Antibody (dG9 (delta G9)), eFluor™ 450, eBioscience™ - FAQs (48-9994-42, 48-9994-41)