IgM Monoclonal Antibody (SA-DA4), eFluor™ 450, eBioscience™ - FAQs
IgM Monoclonal Antibody (SA-DA4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IgM Monoclonal Antibody (SA-DA4), eFluor™ 450, eBioscience™ - FAQs (48-9998-41)