Racks for Matrix™ 2D Barcoded Storage Tubes, 1.4mL, empty latch rack - FAQs
Racks for Matrix™ 2D Barcoded Storage Tubes, 1.4mL, empty latch rack - FAQs
View additional product information for Racks for Matrix™ 2D Barcoded Storage Tubes - FAQs (4905, 4890, 4893, 4896, 4897, 4898, 4899, 4900)
2 product FAQs found
Which racks are suitable for storage of the tubes in which TaqMan assays are delivered?
The Racks for Matrix 2D Barcoded Storage Tubes (Cat. No. 4890) are suitable for storage of narrower tubes. For more products and catalog numbers, see the following link.
Find additional tips, troubleshooting help, and resources within our TaqMan® Primers and Probes Support Center
Can I store the Thermo Scientific 2D barcoded storage tubes in standard 10x10 cryoboxes?
Yes, the Thermo Scientific 2D barcoded storage tubes can be stored in standard cryoboxes. However, we do not carry 2D barcode readers capable of reading a full box of 2D barcodes in this storage format.