eBioscience™ Streptavidin eFluor™ 710 Conjugate, 25 μg - FAQs
eBioscience™ Streptavidin eFluor™ 710 Conjugate, 25 μg - FAQs
View additional product information for eBioscience™ Streptavidin eFluor™ 710 Conjugate - FAQs (49-4317-82, 49-4317-80)