eBioscience™ Streptavidin eFluor™ 710 Conjugate
Invitrogen™

eBioscience™ Streptavidin eFluor™ 710 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
49-4317-82100 μg
49-4317-8025 μg
Catalog number 49-4317-82
Price (USD)
208.65
Online exclusive
278.00
Save 69.35
Each
Quantity:
100 μg
Price (USD)
208.65
Online exclusive
278.00
Save 69.35
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Red Laser

Emit

710 nm

Excite

633 - 647 nm

Reported Application

Flow Cytometric Analysis

For Research Use Only
Specifications
Concentration0.2 mg/mL
Label or DyeeFluor 710
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity100 μg
Target MoleculeBiotin
ConjugateeFluor 710
FormLiquid
Product LineeBioscience
Unit SizeEach

Frequently asked questions (FAQs)

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.