CD3 Monoclonal Antibody (17A2), eFluor™ 660, eBioscience™ - FAQs
CD3 Monoclonal Antibody (17A2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (17A2), eFluor™ 660, eBioscience™ - FAQs (50-0032-82, 50-0032-80)