CD3 Monoclonal Antibody (OKT3), eFluor™ 660, eBioscience™ - FAQs
CD3 Monoclonal Antibody (OKT3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (OKT3), eFluor™ 660, eBioscience™ - FAQs (50-0037-41, 50-0037-42)