CD4 Monoclonal Antibody (GK1.5), eFluor™ 660, eBioscience™ - FAQs
CD4 Monoclonal Antibody (GK1.5), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (GK1.5), eFluor™ 660, eBioscience™ - FAQs (50-0041-80, 50-0041-82)