CD4 Monoclonal Antibody (OKT4 (OKT-4)), eFluor™ 660, eBioscience™ - FAQs
CD4 Monoclonal Antibody (OKT4 (OKT-4)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (OKT4 (OKT-4)), eFluor™ 660, eBioscience™ - FAQs (50-0048-41)