CD8a Monoclonal Antibody (53-6.7), eFluor™ 660, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), eFluor™ 660, eBioscience™ - FAQs (50-0081-82, 50-0081-80)