CD11b/c Monoclonal Antibody (OX42), eFluor™ 660, eBioscience™ - FAQs
CD11b/c Monoclonal Antibody (OX42), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD11b/c Monoclonal Antibody (OX42), eFluor™ 660, eBioscience™ - FAQs (50-0110-82, 50-0110-80)