CD11b Monoclonal Antibody (M1/70), eFluor™ 660, eBioscience™ - FAQs
CD11b Monoclonal Antibody (M1/70), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD11b Monoclonal Antibody (M1/70), eFluor™ 660, eBioscience™ - FAQs (50-0112-80, 50-0112-82)