CD11c Monoclonal Antibody (N418), eFluor™ 660, eBioscience™ - FAQs
CD11c Monoclonal Antibody (N418), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (N418), eFluor™ 660, eBioscience™ - FAQs (50-0114-82, 50-0114-80)