CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 660, eBioscience™ - FAQs
CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 660, eBioscience™ - FAQs (50-0193-80, 50-0193-82)