CD20 Monoclonal Antibody (L26), eFluor™ 660, eBioscience™ - FAQs
CD20 Monoclonal Antibody (L26), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (L26), eFluor™ 660, eBioscience™ - FAQs (50-0202-82, 50-0202-80)