CD23 Monoclonal Antibody (B3B4), eFluor™ 660, eBioscience™ - FAQs
CD23 Monoclonal Antibody (B3B4), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD23 Monoclonal Antibody (B3B4), eFluor™ 660, eBioscience™ - FAQs (50-0232-80, 50-0232-82)