CD25 Monoclonal Antibody (eBio7D4 (7D4)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (eBio7D4 (7D4)), eFluor™ 660, eBioscience™ - FAQs (50-0252-80, 50-0252-82)