SIGLEC H Monoclonal Antibody (eBio440c), eFluor™ 660, eBioscience™ - FAQs
SIGLEC H Monoclonal Antibody (eBio440c), eFluor™ 660, eBioscience™ - FAQs
View additional product information for SIGLEC H Monoclonal Antibody (eBio440c), eFluor™ 660, eBioscience™ - FAQs (50-0333-80)