CD36 Monoclonal Antibody (eBioNL07 (NL07)), eFluor™ 660, eBioscience™ - FAQs
CD36 Monoclonal Antibody (eBioNL07 (NL07)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD36 Monoclonal Antibody (eBioNL07 (NL07)), eFluor™ 660, eBioscience™ - FAQs (50-0369-42, 50-0369-41)