CD38 Monoclonal Antibody (14.27), eFluor™ 660, eBioscience™ - FAQs
CD38 Monoclonal Antibody (14.27), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (14.27), eFluor™ 660, eBioscience™ - FAQs (50-0380-80)