CD39 Monoclonal Antibody (24DMS1), eFluor™ 660, eBioscience™ - FAQs
CD39 Monoclonal Antibody (24DMS1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD39 Monoclonal Antibody (24DMS1), eFluor™ 660, eBioscience™ - FAQs (50-0391-80)