CD39 Monoclonal Antibody (24DMS1), eFluor™ 660, eBioscience™ - FAQs
CD39 Monoclonal Antibody (24DMS1), eFluor™ 660, eBioscience™ - FAQs
查看更多产品信息 CD39 Monoclonal Antibody (24DMS1), eFluor™ 660, eBioscience™ - FAQs (50-0391-82)
5 个常见问题解答