LYVE1 Monoclonal Antibody (ALY7), eFluor™ 660, eBioscience™ - FAQs
LYVE1 Monoclonal Antibody (ALY7), eFluor™ 660, eBioscience™ - FAQs
View additional product information for LYVE1 Monoclonal Antibody (ALY7), eFluor™ 660, eBioscience™ - FAQs (50-0443-80, 50-0443-82)