CD45R (B220) Monoclonal Antibody (RA3-6B2), eFluor™ 660, eBioscience™ - FAQs
CD45R (B220) Monoclonal Antibody (RA3-6B2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD45R (B220) Monoclonal Antibody (RA3-6B2), eFluor™ 660, eBioscience™ - FAQs (50-0452-82, 50-0452-80)