CD56 (NCAM) Monoclonal Antibody (5tukon56), eFluor™ 660, eBioscience™ - FAQs
CD56 (NCAM) Monoclonal Antibody (5tukon56), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD56 (NCAM) Monoclonal Antibody (5tukon56), eFluor™ 660, eBioscience™ - FAQs (50-0565-80)