CD62L (L-Selectin) Monoclonal Antibody (OX85), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD62L (L-Selectin) Monoclonal Antibody (OX85), eFluor™ 660, eBioscience™ - FAQs (50-0623-82, 50-0623-80)