CD63 Monoclonal Antibody (H5C6), eFluor™ 660, eBioscience™ - FAQs
CD63 Monoclonal Antibody (H5C6), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD63 Monoclonal Antibody (H5C6), eFluor™ 660, eBioscience™ - FAQs (50-0639-41, 50-0639-42)