CD68 Monoclonal Antibody (FA-11), eFluor™ 660, eBioscience™ - FAQs
CD68 Monoclonal Antibody (FA-11), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD68 Monoclonal Antibody (FA-11), eFluor™ 660, eBioscience™ - FAQs (50-0681-82, 50-0681-80)