CD70 Monoclonal Antibody (113-16), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD70 Monoclonal Antibody (113-16), eFluor™ 660, eBioscience™ - FAQs (50-0709-41, 50-0709-42)