CD72 Monoclonal Antibody (J4-117), eFluor™ 660, eBioscience™ - FAQs
CD72 Monoclonal Antibody (J4-117), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD72 Monoclonal Antibody (J4-117), eFluor™ 660, eBioscience™ - FAQs (50-0729-42)