CD75 Monoclonal Antibody (LN-1), eFluor™ 660, eBioscience™ - FAQs
CD75 Monoclonal Antibody (LN-1), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD75 Monoclonal Antibody (LN-1), eFluor™ 660, eBioscience™ - FAQs (50-0759-41, 50-0759-42)