CD79a Monoclonal Antibody (24C2.5), eFluor™ 660, eBioscience™ - FAQs
CD79a Monoclonal Antibody (24C2.5), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD79a Monoclonal Antibody (24C2.5), eFluor™ 660, eBioscience™ - FAQs (50-0791-80, 50-0791-82)