CD83 Monoclonal Antibody (Michel-17 (Michel17)), eFluor™ 660, eBioscience™ - FAQs
CD83 Monoclonal Antibody (Michel-17 (Michel17)), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD83 Monoclonal Antibody (Michel-17 (Michel17)), eFluor™ 660, eBioscience™ - FAQs (50-0831-82, 50-0831-80)