CD106 (VCAM-1) Monoclonal Antibody (429), eFluor™ 660, eBioscience™ - FAQs
CD106 (VCAM-1) Monoclonal Antibody (429), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD106 (VCAM-1) Monoclonal Antibody (429), eFluor™ 660, eBioscience™ - FAQs (50-1061-82, 50-1061-80)