CD127 Monoclonal Antibody (A7R34), eFluor™ 660, eBioscience™ - FAQs
CD127 Monoclonal Antibody (A7R34), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (A7R34), eFluor™ 660, eBioscience™ - FAQs (50-1271-82, 50-1271-80)