CD146 Monoclonal Antibody (P1H12), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD146 Monoclonal Antibody (P1H12), eFluor™ 660, eBioscience™ - FAQs (50-1469-42, 50-1469-41)