CD152 (CTLA-4) Monoclonal Antibody (14D3), eFluor™ 660, eBioscience™ - FAQs
CD152 (CTLA-4) Monoclonal Antibody (14D3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD152 (CTLA-4) Monoclonal Antibody (14D3), eFluor™ 660, eBioscience™ - FAQs (50-1529-42)