CD160 Monoclonal Antibody (BY55), eFluor™ 660, eBioscience™ - FAQs
CD160 Monoclonal Antibody (BY55), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD160 Monoclonal Antibody (BY55), eFluor™ 660, eBioscience™ - FAQs (50-1609-41, 50-1609-42)